3-Amino-2-chloropyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Chloro-3-pyridylamine, 2-Chloropyridin-3-amine

214320-10G 214320-250G 214320-50G
BT214320-10GEA 34 USD
BT214320-10G BT214320-250G BT214320-50G
3-Amino-2-chloropyridine ≥98%
3-Amino-2-chloropyridine

CAS: 6298-19-7; EC No: 228-572-9; MDL No: MFCD00006238 Crystalline; Molecular Formula: C5H5ClN2; MW: 128.56 Melting Point: 76-78°


  • Crystalline

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 76…78 °C
MDL Number: MFCD00006238
CAS Number: 6298-19-7
EINECS: 228-572-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR