1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers) ≥97%
Supplier: BeanTown Chemical
Synonyms:
Hexahydroisobenzofuran-1,3-dione, Cyclohexane-1,2-dicarboxylic anhydride
214310-100G
214310-500G
BT214310-100GEA
39
USD
BT214310-100G
BT214310-500G
1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers) ≥97%
1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers)
CAS: 85-42-7; EC No: 201-604-9; MDL No: MFCD00064863
Solid; Molecular Formula: C8H10O3 ; MW: 154.16
Melting Point: 32-34°; Boiling Point: 158°/17 mmHg
Moisture Sensitive
- Moisture Sensitive
- Solid
Formula:
C₈H₁₀O₃ MW: 154.17 g/mol Boiling Pt: 158 °C/17 mmHg Melting Pt: 32…34 °C |
MDL Number:
MFCD00064863 CAS Number: 85-42-7 EINECS: 201-604-9 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...