Rhodium(III) chloride hydrate

Supplier: BeanTown Chemical

Synonyms: Rhodium trichloride hydrate

214270-1G 214270-250MG 214270-5G
BT214270-1GEA 436 USD
BT214270-1G BT214270-250MG BT214270-5G
Rhodium(III) chloride hydrate
Rhodium (III) chloride hydrate

CAS: 20765-98-4; MDL No: MFCD00149839 Crystalline; Linear Formula: RhCl3·xH2O; MW: 209.26 (anhydrous) Melting Point: 100° (decomposes) Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: Cl₃Rh·H₂O
MW: 227.28 g/mol
Melting Pt: 100 °C (dec.)
MDL Number: MFCD00149839
CAS Number: 20765-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR