4-Methyl-1H-pyrazole ≥97%
Supplier: BeanTown Chemical
CAS: 7554-65-6; EC No: 231-445-0; MDL No: MFCD00005245; RTECS: UQ7350000
Liquid; Molecular Formula: C4H6N2; MW: 82.10
Boiling Point: 99-100°/6 mmHg; Flash point: 96°C (205°F)
Density (g/mL): 0.993; Refractive Index: 1.495
Hygroscopic
- RTECS: UQ7350000
- Refractive index: 1.495
- Hygroscopic
- Liquid
Formula:
C₄H₆N₂ Boiling Pt: 99…100 °C/6 mmHg Density: 0.993 g/mL Flash Pt: 96 °C (205 °F) |
MDL Number:
MFCD00005245 CAS Number: 7554-65-6 EINECS: 231-445-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...