Lanthanum(III) carbonate hydrate ≥99.99%
Supplier: BeanTown Chemical
214225-250G
214225-50G
BT214225-250GEA
321
USD
BT214225-250G
BT214225-50G
Lanthanum(III) carbonate hydrate ≥99.99%
Lanthanum(III) carbonate hydrate
CAS: 54451-24-0; EC No: 209-599-5; MDL No: MFCD00011069
Powder/Lump; Linear Formula: La2(CO3)3·xH2O; MW: 457.85 (anhydrous)
Hygroscopic
- Hygroscopic
- Powder/Lump
Formula:
La₂(CO₃)₃.xH₂O MW: 457.85 g/mol (anhy) |
MDL Number:
MFCD00011069 CAS Number: 54451-24-0 EINECS: 209-599-5 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...