Resazurin sodium salt

Supplier: BeanTown Chemical

Synonyms: 7-Sodiooxy-3H-phenoxazin-3-one 10-oxide, Sodium 7-oxido-3-oxo-3H-phenoxazine 10-oxide, Resazurin sodium, Resazurin

214215-1G 214215-5G
BT214215-1GEA 38.13 USD
BT214215-1G BT214215-5G
Resazurin sodium salt
Resazurin sodium salt

CAS: 62758-13-8; EC No: 263-718-5; MDL No: MFCD00005036; RTECS: SP7700000 Powder; Molecular Formula: C12H6NNaO4; MW: 251.18


  • RTECS: SP7700000
  • Powder

Formula: C₁₂H₆NNaO₄
MW: 251.17 g/mol
MDL Number: MFCD00005036
CAS Number: 62758-13-8
EINECS: 263-718-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR