Benzyl cyanoacetate ≥97%

Supplier: BeanTown Chemical

Synonyms: Cyanoacetic benzyl ester, Phenylmethyl cyanoacetate

214200-100G 214200-25G
BT214200-100GEA 480 USD
BT214200-100G BT214200-25G
Benzyl cyanoacetate ≥97%
Benzyl cyanoacetate

CAS: 14447-18-8; MDL No: MFCD00051719 Liquid; Molecular Formula: C10H9NO2; MW: 175.18 Boiling Point: 115-125°/0.3 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.144; Refractive Index: 1.520


  • Refractive index: 1.520
  • Liquid

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 115…125 °C/0.3 mmHg
Density: 1.144 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00051719
CAS Number: 14447-18-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR