Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate ≥98%

Supplier: BeanTown Chemical

Synonyms: Hantzsch Ester, 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Diethyl Ester, Diethyl 1,4-dihydro-3,5-pyridinecarboxylate, Etidin, Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate

214195-1G 214195-250MG 214195-5G
BT214195-1GEA 67 USD
BT214195-1G BT214195-250MG BT214195-5G
Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate ≥98%
Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate

CAS: 1149-23-1; EC No: 214-561-6; MDL No: MFCD00005951; RTECS: US7970100 Solid; Molecular Formula: C13H19NO4; MW: 253.29 Melting Point: 178-183°


  • RTECS: US7970100
  • Solid

Formula: C₁₃H₁₉NO₄
MW: 253.30 g/mol
Melting Pt: 178…183 °C
MDL Number: MFCD00005951
CAS Number: 1149-23-1
EINECS: 214-561-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR