Tetraammineplatinum(II) chloride monohydrate ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Tetraammineplatinum (II) chloride hydrate
214170-1G
214170-5G
BT214170-1GEA
189.88
USD
BT214170-1G
BT214170-5G
Tetraammineplatinum(II) chloride monohydrate ≥99.995% (trace metals basis)
Tetraammineplatinum(II) chloride monohydrate
CAS: 13933-33-0; EC No: 237-706-5; MDL No: MFCD00149947
Crystalline; Linear Formula: Pt(NH3)4Cl2·H2O; MW: 352.13
Melting Point: 250° (decomposes)
Density (g/mL): 2.74
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
Cl₂H₁₄N₄OPt MW: 352.123 g/mol Melting Pt: 250 °C (dec.) Density: 2.74 g/mL |
MDL Number:
MFCD00149947 CAS Number: 13933-33-0 EINECS: 237-706-5 |
Specification Test Results
Assay | 99.995% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...