3,5-Di-tert-butylsalicylic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 3,5-Bis-tert-butylsalicylic acid, 3,5-Di-tert-butyl-2-hydroxybenzoic Acid

214155-25G 214155-5G
BT214155-25GEA 115 USD
BT214155-25G BT214155-5G
3,5-Di-tert-butylsalicylic acid ≥97%
3,5-Di-tert-butylsalicylic acid

CAS: 19715-19-6; EC No: 243-246-6; MDL No: MFCD00059058 Powder; Molecular Formula: C15H22O4; MW: 250.33 Melting Point: 157-162°; Flash point: 179°C (354°F)


  • Powder

Formula: C₁₅H₂₂O₃
MW: 250.34 g/mol
Melting Pt: 157…162 °C
Flash Pt: 179 °C (354 °F)
MDL Number: MFCD00059058
CAS Number: 19715-19-6
EINECS: 243-246-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR