Ammonium hydrogen carbonate ≥99%

Supplier: BeanTown Chemical

Synonyms: Ammonium bicarbonate

214150-250G 214150-50G
BT214150-250GEA 222.94 USD
BT214150-250G BT214150-50G
Ammonium hydrogen carbonate ≥99%
Ammonium hydrogen carbonate

CAS: 1066-33-7; EC No: 213-911-5; MDL No: MFCD00012138; RTECS: BO8600000 Powder; Linear Formula: NH4HCO3; MW: 79.06 Density (g/mL): 1.586


  • RTECS: BO8600000
  • Powder

Formula: NH₄HCO₃
MW: 79.06 g/mol
Melting Pt: 35
Density: 1.586 g/mL
MDL Number: MFCD00012138
CAS Number: 1066-33-7
EINECS: 213-911-5
Merck Index: 13,00497

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR