1-Decanethiol ≥96%

Supplier: BeanTown Chemical

Synonyms: n-Decanethiol, 1-Mercaptodecane, n-Decyl mercaptan

214140-100ML 214140-25ML 214140-500ML
BT214140-100MLEA 72 USD
BT214140-100ML BT214140-25ML BT214140-500ML
1-Decanethiol ≥96%
1-Decanethiol

CAS: 143-10-2; EC No: 205-584-2; MDL No: MFCD00004884 Liquid; Linear Formula: CH3(CH2)9SH; Molecular Formula: C10H22S; MW: 174.35 Melting Point: -26°; Boiling Point: 114°/13 mmHg; Flash point: 98°C (208°F) Density (g/mL): 0.824; Refractive Index: 1.457 Air Sensitive


  • Refractive index: 1.457
  • Air Sensitive
  • Liquid

Formula: C₁₀H₂₂S
MW: 174.35 g/mol
Boiling Pt: 114 °C/13 mmHg
Melting Pt: –26 °C
Density: 0.824 g/mL
Flash Pt: 98 °C (208 °F)
MDL Number: MFCD00004884
CAS Number: 143-10-2
EINECS: 205-584-2
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR