Methyl-3-(3-pyridyl)propionate ≥98%
Supplier: BeanTown Chemical
214100-1G
214100-5G
BT214100-1GEA
41
USD
BT214100-1G
BT214100-5G
Methyl-3-(3-pyridyl)propionate ≥98%
Methyl-3-(3-pyridyl)propionate
CAS: 84199-98-4; EC No: 000-000-0; MDL No: MFCD01075660
Liquid; Molecular Formula: C9H11NO2; MW: 165.19
Boiling Point: 134°/12 mmHg
Refractive Index: 1.501
- Refractive index: 1.501
- Liquid
Formula:
C₉H₁₁NO₂ Boiling Pt: 134 °C/12 mmHg |
MDL Number:
MFCD01075660 CAS Number: 84199-98-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...