(S)-2-Hydroxy-3-phenylpropanoic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: L-β-Phenyllactic acid, (S)-(-)-3-Phenyllactic Acid, L-3-Phenyllactic Acid, L(-)-3-Phenyllactic acid, (S)-(-)-2-Hydroxy-3-phenylpropionic Acid, L-(-)-3-Phenyllactic acid

214035-1G 214035-25G 214035-5G
BT214035-1GEA 29 USD
BT214035-1G BT214035-25G BT214035-5G
(S)-2-Hydroxy-3-phenylpropanoic acid ≥98%
(S)-2-Hydroxy-3-phenylpropanoic acid

CAS: 20312-36-1; EC No: 243-726-5; MDL No: MFCD00004244 Crystalline; Molecular Formula: C9H10O3 ; MW: 166.17 Melting Point: 122-124° Optical Rotation: [α]24/D -20.8°, c = 2 in water


  • Optical rotation: [α]24/D -20.8°, c = 2 in water
  • Crystalline

Formula: C₉H₁₀O₃
MW: 166.17 g/mol
Melting Pt: 122…124 °C
MDL Number: MFCD00004244
CAS Number: 20312-36-1
EINECS: 243-726-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR