1,2-Bis(diphenylphosphino)ethane ≥97%
Supplier: BeanTown Chemical
Synonyms:
Ethylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)ethane, DIPHOS, 1,2-Bis(diphenylphosphino)ethane, Ethane-1,3-diylbis(diphenylphosphine)
214010-100G
214010-25G
214010-5G
BT214010-100GEA
419
USD
BT214010-100G
BT214010-25G
BT214010-5G
1,2-Bis(diphenylphosphino)ethane ≥97%
1,2-Bis(diphenylphosphino)ethane
CAS: 1663-45-2; EC No: 216-769-2; MDL No: MFCD00003047
Powder; Molecular Formula: C26H24P2; MW: 398.42
Melting Point: 137-142°
Air Sensitive
- Air Sensitive
- Powder
Formula:
C₂₆H₂₄P₂ Melting Pt: 137…142 °C |
MDL Number:
MFCD00003047 CAS Number: 1663-45-2 EINECS: 216-769-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...