7-Amino-4-methylcoumarin ≥98%

Supplier: BeanTown Chemical

Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC

213935-1G 213935-250MG 213935-5G
BT213935-1GEA 115.4 USD
BT213935-1G BT213935-250MG BT213935-5G
7-Amino-4-methylcoumarin ≥98%
7-Amino-4-methylcoumarin

CAS: 26093-31-2; EC No: 247-454-8; MDL No: MFCD00006868 Solid; Molecular Formula: C10H9NO2; MW: 175.18 Melting Point: 223-226° Light Sensitive


  • Light Sensitive
  • Solid

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 223…226 °C
MDL Number: MFCD00006868
CAS Number: 26093-31-2
EINECS: 247-454-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR