Benzofuran-2-carboxaldehyde ≥99%
Supplier: BeanTown Chemical
213915-1G
213915-5G
BT213915-1GEA
50
USD
BT213915-1G
BT213915-5G
Benzofuran-2-carboxaldehyde ≥99%
Benzofuran-2-carboxaldehyde
CAS: 4265-16-1; EC No: 224-248-6; MDL No: MFCD00015463
Liquid; Molecular Formula: C9H6O2; MW: 146.14
Boiling Point: 135°/18 mmHg
Density (g/mL): 1.206; Refractive Index: 1.633
Air Sensitive
- Refractive index: 1.633
- Air Sensitive
- Liquid
Formula:
C₉H₆O₂ Boiling Pt: 135 °C/18 mmHg Density: 1.206 g/mL |
MDL Number:
MFCD00015463 CAS Number: 4265-16-1 EINECS: 224-248-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...