L(-)-Cystine ≥99%

Supplier: BeanTown Chemical

213890-1KG 213890-250G 213890-50G
BT213890-1KGEA 341 USD
BT213890-1KG BT213890-250G BT213890-50G
L(-)-Cystine ≥99%
L(-)-Cystine

CAS: 56-89-3; EC No: 200-296-3; MDL No: MFCD00064228; RTECS: HA2690000 Powder; Molecular Formula: C6H12N2O4S2; MW: 240.30 Melting Point: 240° (decomposes) Optical Rotation: [α]20/D -219±5°, c = 1% in 1M HCl


  • RTECS: HA2690000
  • Optical rotation: [α]20/D -219±5°, c = 1% in 1M HCl
  • Powder

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR