Quinine hemisulfate monohydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: Quinine sulphate, Quinine sulphate (2:1) (salt) dihydrate

213810-10G 213810-250G 213810-50G
BT213810-10GEA 38.73 USD
BT213810-10G BT213810-250G BT213810-50G
Quinine hemisulfate monohydrate ≥99%
Quinine hemisulfate monohydrate

CAS: 6119-70-6; EC No: 212-359-2; MDL No: MFCD00150792; RTECS: VA8440000 Powder; Molecular Formula: C20H24N2O2·0.5H2SO4·H2O; MW: 391.47 Melting Point: 225° (decomposes) Optical Rotation: [α]20/D -230±5°, c = 2% in 0.1M HCl Light Sensitive


  • RTECS: VA8440000
  • Optical rotation: [α]20/D -230±5°, c = 2% in 0.1M HCl
  • Light Sensitive
  • Powder

Formula: C₂₀H₂₄N₂O₂·0.5H₂SO₄·H₂O
MW: 391.47 g/mol (373.46 anhy)
Melting Pt: 225 °C (dec.)
MDL Number: MFCD00150792
CAS Number: 6119-70-6
EINECS: 212-359-2

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR