Disodium hexahydroxoplatinate ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Disodium platinate, Sodium hexahydroxyplatinate(IV)

213770-1G 213770-5G
BT213770-1GEA 268 USD
BT213770-1G BT213770-5G
Disodium hexahydroxoplatinate ≥99.9% (trace metals basis)
Disodium hexahydroxoplatinate

CAS: 12325-31-4; EC No: 235-590-0; MDL No: MFCD00014236; RTECS: TP1756000 Crystalline/Powder; Linear Formula: Na2Pt(OH)6; MW: 343.10 Melting Point: 150° (decomposes)


  • RTECS: TP1756000
  • Crystalline/Powder

Formula: H₆Na₂O₆Pt
MW: 343.1 g/mol
Melting Pt: 150 °C (dec.)
MDL Number: MFCD00014236
CAS Number: 12325-31-4
EINECS: 235-590-0

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR