Molybdenum ≥99.95% (trace metals basis), plate, Thickness 12.5 mm

Supplier: BeanTown Chemical

213690-50X200 213690-50X50MM
BT213690-50X20EA 968 USD
BT213690-50X20 BT213690-50X50
Molybdenum ≥99.95% (trace metals basis), plate, Thickness 12.5 mm
Molybdenum

CAS: 7439-98-7; EC No: 231-107-2; MDL No: MFCD00003465 Solid; Molecular Formula: Mo; MW: 95.94 Melting Point: 2617°; Boiling Point: 4612° Density (g/mL): 10.3


  • Solid

Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 4612 °C
Melting Pt: 2617 °C
Density: 10.3 g/mL
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2

Order Now

Specification Test Results

Assay 99.95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR