8-Amino-1-naphthalenesulfonic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 8-Aminonaphthalene-1-sulphonic acid

213685-100G 213685-25G 213685-500G
BT213685-100GEA 71 USD
BT213685-100G BT213685-25G BT213685-500G
8-Amino-1-naphthalenesulfonic acid ≥98%
8-Amino-1-naphthalenesulphonic acid

CAS: 82-75-7; EC No: 201-437-1; MDL No: MFCD00035730 Powder; Molecular Formula: C10H9NO3S; MW: 223.25 Melting Point: 300°


  • Powder

Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00035730
CAS Number: 82-75-7
EINECS: 201-437-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR