Palladium ≥99.9% (trace metals basis), wire Ø 0.406 mm as drawn

Supplier: BeanTown Chemical

213650-1M 213650-5M
BT213650-1MEA 303 USD
BT213650-1M BT213650-5M
Palladium ≥99.9% (trace metals basis), wire Ø 0.406 mm as drawn
Palladium

CAS: 7440-05-3; EC No: 231-115-6; MDL No: MFCD00011167; RTECS: RT3480500 Solid; Molecular Formula: Pd; MW: 106.42 Melting Point: 1554°; Boiling Point: 2970° Density (g/mL): 12.02


  • RTECS: RT3480500
  • Solid

Formula: Pd
Boiling Pt: 2970 °C
Melting Pt: 1554 °C
Density: 12.02 g/mL
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR