Zirconium(IV) acetylacetonate

Supplier: BeanTown Chemical

213610-250G 213610-50G
BT213610-250GEA 98 USD
BT213610-250G BT213610-50G
Zirconium(IV) acetylacetonate
Zirconium(IV) acetylacetonate

CAS: 17501-44-9; EC No: 241-510-5; MDL No: MFCD00000036 Crystalline; Linear Formula: Zr(C5H7O2)4; Molecular Formula: C20H28O8Zr; MW: 487.66 Melting Point: 191-195° Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₂₀H₂₈O₈Zr
Melting Pt: 191…195 °C
MDL Number: MFCD00000036
CAS Number: 17501-44-9
EINECS: 241-510-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR