Picolinamidine hydrochloride ≥97%

Supplier: BeanTown Chemical

213605-1G 213605-25G 213605-5G
BT213605-1GEA 69 USD
BT213605-1G BT213605-25G BT213605-5G
Picolinamidine hydrochloride ≥97%
Picolinamidine hydrochloride

CAS: 51285-26-8; MDL No: MFCD00052271 Solid; Molecular Formula: C6H7N3·HCl; MW: 157.60 Hygroscopic


  • Hygroscopic
  • Solid

Formula: C₆H₈ClN₃
MDL Number: MFCD00052271
CAS Number: 51285-26-8

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR