Tantalum ≥99.95% (trace metals basis), foil, annealed, Thickness 0.127 mm
Supplier: BeanTown Chemical
213555-100X100
213555-200X200
213555-50X50MM
BT213555-100X1EA
196
USD
BT213555-100X1
BT213555-200X2
BT213555-50X50
Tantalum ≥99.95% (trace metals basis), foil, annealed, Thickness 0.127 mm
Tantalum
CAS: 7440-25-7; EC No: 231-135-5; MDL No: MFCD00011252; RTECS: WW5505000
Solid; Molecular Formula: Ta; MW: 180.95
Melting Point: 2996°; Boiling Point: 5425°
Density (g/mL): 16.69
- RTECS: WW5505000
- Solid
Formula:
Ta Boiling Pt: 5425 °C Melting Pt: 2996 °C Density: 16.69 g/mL |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...