2-Hydroxyisobutyric acid, (max. 2% H₂O) ≥99% (dry weight)
Supplier: BeanTown Chemical
Synonyms:
2-Hydroxy-2-methylpropionic acid, α-Hydroxyisobutyric acid, 2-Hydroxy-2-methylpropanoic acid
213545-100G
213545-25G
213545-500G
BT213545-100GEA
92.32
USD
BT213545-100G
BT213545-25G
BT213545-500G
2-Hydroxyisobutyric acid, (max. 2% H₂O) ≥99% (dry weight)
2-Hydroxyisobutyric acid
CAS: 594-61-6; EC No: 209-848-8; MDL No: MFCD00004459
Crystalline; Molecular Formula: C4H8O3; MW: 104.10
Melting Point: 76-80°; Boiling Point: 84°/1.5 mmHg
- Crystalline
Formula:
C₄H₈O₃ MW: 104.11 g/mol Boiling Pt: 84 °C/1.5 mmHg Melting Pt: 76…80 °C |
MDL Number:
MFCD00004459 CAS Number: 594-61-6 EINECS: 209-848-8 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...