Tetrachloro-p-benzoquinone ≥97%

Supplier: BeanTown Chemical

Synonyms: Tetrachloroquinone, 2,3,5,6-Tetrachloro-1,4-benzoquinone, 4-Chloranil, p-Chloranil, 2,3,5,6-Tetrachlorobenzo-1,4-quinone, Tetrachloro-1,4-benzoquinone

213530-100G 213530-25G 213530-500G
BT213530-100GEA 92 USD
BT213530-100G BT213530-25G BT213530-500G
Tetrachloro-p-benzoquinone ≥97%
Tetrachloro-p-benzoquinone

CAS: 118-75-2; EC No: 204-274-4; MDL No: MFCD00001594; RTECS: DK6825000 Crystalline; Linear Formula: C6Cl4(=O)2; MW: 245.88 Melting Point: 289° (decomposes) Density (g/mL): 1.97 Light Sensitive


  • RTECS: DK6825000
  • Light Sensitive
  • Crystalline

Formula: C₆Cl₄O₂
MW: 245.88 g/mol
Melting Pt: 289 °C (dec.)
Density: 1.97 g/mL
MDL Number: MFCD00001594
CAS Number: 118-75-2
EINECS: 204-274-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR