Strontium sulphate

Supplier: BeanTown Chemical

213455-100G 213455-25G
BT213455-100GEA 52 USD
BT213455-100G BT213455-25G
Strontium sulphate
Strontium sulphate

CAS: 7759-02-6; EC No: 231-850-2; MDL No: MFCD00036274 Crystalline; Linear Formula: SrSO4; MW: 183.68 Melting Point: 1600° Density (g/mL): 3.960


  • Crystalline

Formula: O₄SSr
Melting Pt: 1600 °C
Density: 3.960 g/mL
MDL Number: MFCD00036274
CAS Number: 7759-02-6
EINECS: 231-850-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR