Carmine

Supplier: BeanTown Chemical

Synonyms: Carminic acid, Cochineal, Alum lake of Carminic Acid, Natural Red 4, FD and C Carmine, CI 75470, Carmine, calcium/aluminum lake

213410-25G 213410-5G
BT213410-25GEA 129.74 USD
BT213410-25G BT213410-5G
Carmine
Carmine

CAS: 1390-65-4; EC No: 215-724-4; MDL No: MFCD00167028; RTECS: FH8891000 Powder


  • RTECS: FH8891000
  • Powder

Formula: C₂₂H₂₀O₁₃
MW: 492.389 g/mol
MDL Number: MFCD00167028
CAS Number: 1390-65-4
EINECS: 215-724-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR