6-Fluorosalicylaldehyde ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Fluoro-6-hydroxybenzaldehyde

213395-1G 213395-250MG
BT213395-1GEA 257 USD
BT213395-1G BT213395-250MG
6-Fluorosalicylaldehyde ≥97%
6-Fluorosalicylaldehyde

CAS: 38226-10-7; MDL No: MFCD01090996 Solid; Molecular Formula: C7H5FO2; MW: 140.12 Melting Point: 37-39° Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₇H₅FO₂
MW: 140.11 g/mol
Melting Pt: 37…39 °C
MDL Number: MFCD01090996
CAS Number: 38226-10-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR