2,4,6-Tribromotoluene ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-Methyl-2,4,6-tribromobenzene, 1,3,5-Tribromo-2-methylbenzene

213390-10G 213390-50G
BT213390-10GEA 60 USD
BT213390-10G BT213390-50G
2,4,6-Tribromotoluene ≥98%
2,4,6-Tribromotoluene

CAS: 6320-40-7; EC No: 228-672-2; MDL No: MFCD00013527 Powder; Molecular Formula: C7H5Br3; MW: 328.83 Density (g/mL): 2.48


  • Powder

Formula: C₇H₅Br₃
MW: 328.83 g/mol
Density: 2.48 g/mL
MDL Number: MFCD00013527
CAS Number: 6320-40-7
EINECS: 228-672-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR