2-Chloroimidazole ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Chloro-1H-imidazole

213380-1G 213380-250MG 213380-5G
BT213380-1GEA 249 USD
BT213380-1G BT213380-250MG BT213380-5G
2-Chloroimidazole ≥97%
2-Chloroimidazole

CAS: 16265-04-6; MDL No: MFCD02179530 Solid; Molecular Formula: C3H3ClN2; MW: 102.52 Melting Point: 165-170°


  • Solid

Formula: C₃H₃ClN₂
MW: 102.52 g/mol
Melting Pt: 165…170 °C
MDL Number: MFCD02179530
CAS Number: 16265-04-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR