Methyl-4-iodsalicylate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Methyl 2-hydroxy-4-iodobenzoate
213365-25G
213365-5G
BT213365-25GEA
134
USD
BT213365-25G
BT213365-5G
Methyl-4-iodsalicylate ≥98%
Methyl 4-iodsalicylate
CAS: 18179-39-0; MDL No: MFCD06797864
Solid; Molecular Formula: C8H7IO3; MW: 278.05
Melting Point: 69-73°
Light Sensitive
- Light Sensitive
- Solid
Formula:
C₈H₇IO₃ Melting Pt: 69…73 °C |
MDL Number:
MFCD06797864 CAS Number: 18179-39-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...