4,4'-Dimethylbiphenyl ≥99%

Supplier: BeanTown Chemical

213345-1G 213345-25G 213345-5G
BT213345-1GEA 32 USD
BT213345-1G BT213345-25G BT213345-5G
4,4'-Dimethylbiphenyl ≥99%
4,4'-Dimethylbiphenyl

CAS: 613-33-2; EC No: 210-337-7; MDL No: MFCD00008545 Crystalline; Molecular Formula: C14H14; MW: 182.26 Melting Point: 118-120°; Boiling Point: 295°


  • Crystalline

Formula: C₁₄H₁₄
Boiling Pt: 295 °C
Melting Pt: 118…120 °C
MDL Number: MFCD00008545
CAS Number: 613-33-2
EINECS: 210-337-7

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR