Salbutamol ≥99%
Supplier: BeanTown Chemical
Synonyms:
Proventil, Albuterol
213280-1G
213280-250MG
213280-5G
BT213280-1GEA
98
USD
BT213280-1G
BT213280-250MG
BT213280-5G
Salbutamol ≥99%
Salbutamol
CAS: 18559-94-9; EC No: 242-424-0; MDL No: MFCD00148978; RTECS: ZE4400000
Powder; Molecular Formula: C13H21NO3; MW: 239.31
- RTECS: ZE4400000
- Powder
Formula:
C₁₃H₂₁NO₃ MW: 239.315 g/mol |
MDL Number:
MFCD00148978 CAS Number: 18559-94-9 EINECS: 242-424-0 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...