Bismuth germanium oxide ≥99.9995% (trace metals basis)

Supplier: BeanTown Chemical

213220-100G 213220-25G
BT213220-100GEA 457 USD
BT213220-100G BT213220-25G
Bismuth germanium oxide ≥99.9995% (trace metals basis)
Bismuth germanium oxide

CAS: 12233-56-6; EC No: 235-457-7; MDL No: MFCD00064629 Powder; Molecular Formula: Bi4Ge3O12; MW: 1245.68


  • Powder

Formula: Bi₄Ge₃O₁₂
MW: 1245.68 g/mol
MDL Number: MFCD00064629
CAS Number: 12233-56-6
EINECS: 235-457-7

Order Now

Specification Test Results

Assay 99.9995%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR