L-Proline-tert-butyl ester hydrochloride ≥98%

Supplier: BeanTown Chemical

Synonyms: H-Pro-OtBu.HCl

213210-1G 213210-5G
BT213210-1GEA 57 USD
BT213210-1G BT213210-5G
L-Proline-tert-butyl ester hydrochloride ≥98%
L-Proline-tert-butyl ester hydrochloride

CAS: 5497-76-7; EC No: 226-835-2; MDL No: MFCD00153459 Solid; Molecular Formula: C9H17NO2·HCl; MW: 207.7


  • Solid

Formula: C₉H₁₇NO₂
MW: 207.70 g/mol
MDL Number: MFCD00153459
CAS Number: 5497-76-7
EINECS: 226-835-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR