1,3-Dimethoxybenzene ≥98%
Supplier: BeanTown Chemical
Synonyms:
Resorcinol dimethyl ether
213175-100G
213175-500G
BT213175-100GEA
36.84
USD
BT213175-100G
BT213175-500G
1,3-Dimethoxybenzene ≥98%
1,3-Dimethoxybenzene
CAS: 151-10-0; EC No: 205-783-4; MDL No: MFCD00008384; RTECS: CZ6474000
Liquid; Molecular Formula: C8H10O2; MW: 138.16
Boiling Point: 85-87°/7 mmHg
Density (g/mL): 1.065; Refractive Index: 1.525
- RTECS: CZ6474000
- Refractive index: 1.525
- Liquid
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 85…87 °C/7 mmHg Density: 1.065 g/mL |
MDL Number:
MFCD00008384 CAS Number: 151-10-0 EINECS: 205-783-4 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...