Calcium chloride hexahydrate ≥99.999% (trace metals basis)
Supplier: BeanTown Chemical
213165-1G
213165-25G
213165-5G
BT213165-1GEA
23
USD
BT213165-1G
BT213165-25G
BT213165-5G
Calcium chloride hexahydrate ≥99.999% (trace metals basis)
Calcium chloride hexahydrate
CAS: 7774-34-7; EC No: 233-140-8; MDL No: MFCD00149616
Crystalline Aggregates; Molecular Formula: CaCl2·xH2O; MW: 110.98 (anhydrous)
Melting Point: 176° (decomposes)
Hygroscopic
- Hygroscopic
- Crystalline Aggregates
Formula:
CaCl₂·6H₂O Melting Pt: 176 °C (dec.) |
MDL Number:
MFCD00149616 CAS Number: 7774-34-7 EINECS: 233-140-8 |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...