2,5-Dibromobenzoic acid ≥98%

Supplier: BeanTown Chemical

213160-25G 213160-5G
BT213160-25GEA 242 USD
BT213160-25G BT213160-5G
2,5-Dibromobenzoic acid ≥98%
2,5-Dibromobenzoic acid

CAS: 610-71-9; EC No: 210-234-7; MDL No: MFCD00016494; RTECS: DG6290000 Solid; Molecular Formula: C7H4Br2O2 ; MW: 279.91 Melting Point: 156-159°


  • RTECS: DG6290000
  • Solid

Formula: C₇H₄Br₂O₂
MW: 279.92 g/mol
Melting Pt: 156…159 °C
MDL Number: MFCD00016494
CAS Number: 610-71-9
EINECS: 210-234-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR