Aluminium oxide activated neutral γ-phase ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
213155-100G
213155-500G
BT213155-100GEA
66.82
USD
BT213155-100G
BT213155-500G
Aluminium oxide activated neutral γ-phase ≥99.9% (trace metals basis)
Aluminium oxide activated neutral
CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424
40 Micron APS Powder, S.A. 200 m2/g; Molecular Formula: Al2O3 ; MW: 101.96
Melting Point: 2040°
- 40 Micron APS Powder, S.A. 200 m²/g
Formula:
Al₂O₃ MW: 101.96 g/mol Melting Pt: 2040 °C |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...