Tetraethylammonium bromide ≥98%

Supplier: BeanTown Chemical

Synonyms: Tetrylammonium bromide

213145-1KG 213145-250G 213145-5KG
BT213145-1KGEA 164.12 USD
BT213145-1KG BT213145-250G BT213145-5KG
Tetraethylammonium bromide ≥98%
Tetraethylammonium bromide

CAS: 71-91-0; EC No: 200-769-4; MDL No: MFCD00011825; RTECS: BS5950000 Crystalline; Linear Formula: (CH3CH2)4N(Br); Molecular Formula: C8H20BrN; MW: 210.16 Melting Point: 285° (decomposes) Density (g/mL): 1.397 Hygroscopic


  • RTECS: BS5950000
  • Hygroscopic
  • Crystalline

Formula: C₈H₂₀BrN
MW: 210.16 g/mol
Melting Pt: 285 °C (dec.)
Density: 1.397 g/mL
MDL Number: MFCD00011825
CAS Number: 71-91-0
EINECS: 200-769-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR