trans,trans-Farnesol ≥97%

Supplier: BeanTown Chemical

Synonyms: (2E,6E)-3,7,11-Trimethyldodeca-2,6,10-trien-1-ol

213130-1G 213130-25G 213130-5G
BT213130-1GEA 24 USD
BT213130-1G BT213130-25G BT213130-5G
trans,trans-Farnesol ≥97%
trans,trans-Farnesol

CAS: 106-28-5; EC No: 225-004-1; MDL No: MFCD00002918; RTECS: JR4979000 Liquid; Molecular Formula: C15H26O; MW: 222.37 Boiling Point: 137°/3 mmHg; Flash point: 96°C (205°F) Density (g/mL): 0.879; Refractive Index: 1.490 Light Sensitive


  • RTECS: JR4979000
  • Refractive index: 1.490
  • Light Sensitive
  • Liquid

Formula: C₁₅H₂₆O
MW: 222.37 g/mol
Boiling Pt: 137 °C/3 mmHg
Density: 0.879 g/mL
Flash Pt: 96 °C (205 °F)
MDL Number: MFCD00002918
CAS Number: 106-28-5
EINECS: 225-004-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR