4-Hydroxy-3-nitropyridine ≥95%

Supplier: BeanTown Chemical

Synonyms: 3-Nitro-4-pyridon, 3-Nitro-4-pyridone, 3-Nitropyridin-4-ol, 3-Nitro-4-pyridinol

213080-100G 213080-25G 213080-5G
BT213080-100GEA 529 USD
BT213080-100G BT213080-25G BT213080-5G
4-Hydroxy-3-nitropyridine ≥95%
4-Hydroxy-3-nitropyridine

CAS: 5435-54-1; MDL No: MFCD00128874 Crystalline/Powder; Molecular Formula: C5H4N2O3 ; MW: 140.10 Melting Point: 285° (decomposes)


  • Crystalline/Powder

Formula: C₅H₄N₂O₃
MW: 140.10 g/mol
Melting Pt: 285 °C (dec.)
MDL Number: MFCD00128874
CAS Number: 5435-54-1

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR