1,4-Phenylenediacetic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
2,2'-(1,4-Phenylene)diacetic acid
213060-1G
213060-25G
213060-5G
BT213060-1GEA
26
USD
BT213060-1G
BT213060-25G
BT213060-5G
1,4-Phenylenediacetic acid ≥97%
1,4-Phenylenediacetic acid
CAS: 7325-46-4; EC No: 230-801-2; MDL No: MFCD00004354
Powder; Molecular Formula: C10H10O4; MW: 194.18
Melting Point: 248-252°
- Powder
Formula:
C₁₀H₁₀O₄ MW: 194.19 g/mol Melting Pt: 248…252 °C |
MDL Number:
MFCD00004354 CAS Number: 7325-46-4 EINECS: 230-801-2 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...