Potassium sorbate ≥99%

Supplier: BeanTown Chemical

Synonyms: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate

213035-1KG 213035-250G 213035-5KG
BT213035-1KGEA 124.2 USD
BT213035-1KG BT213035-250G BT213035-5KG
Potassium sorbate ≥99%
Potassium sorbate

CAS: 24634-61-5; EC No: 246-376-1; MDL No: MFCD00016546; RTECS: WG2170000 Solid; Linear Formula: CH3CH=CHCH=CHCOOK; Molecular Formula: C6H7KO2; MW: 150.22 Melting Point: 210° (decomposes) Density (g/mL): 1.36


  • RTECS: WG2170000
  • Solid

Formula: CH₃CH=CHCH=CHCOOK
MW: 150.22 g/mol
Melting Pt: 210 °C (dec.)
Density: 1.36 g/mL
MDL Number: MFCD00016546
CAS Number: 24634-61-5
EINECS: 246-376-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=500421971592bd8f37c1a6b88f97c1de033cbf87&vid=e8c4a1d0da78946f174af6a94e5c78fc9c16de05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR