Aluminum dihydrogen phosphate 50% (w/w) in water

Supplier: BeanTown Chemical

Synonyms: Aluminum hydrogen phosphate, Aluminium dihydrogen phosphate, Aluminium tris(dihydrogen phosphate), Aluminum phosphate monobasic

213015-1KG 213015-5KG
BT213015-1KGEA 68.16 USD
BT213015-1KG BT213015-5KG
Aluminum dihydrogen phosphate 50% (w/w) in water
Aluminum dihydrogen phosphate

CAS: 13530-50-2; MDL No: MFCD00014152 Liquid; Molecular Formula: AlH6O12P3; MW: 317.94 Density (g/mL): 1.5


  • Liquid

Formula: AlH₆O₁₂P₃
MW: 317.939 g/mol
Density: 1.5 g/mL
MDL Number: MFCD00014152
CAS Number: 13530-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR