9-Fluorenylmethanol ≥99%
Supplier: BeanTown Chemical
213005-100G
213005-25G
213005-5G
BT213005-100GEA
593
USD
BT213005-100G
BT213005-25G
BT213005-5G
9-Fluorenylmethanol ≥99%
9-Fluorenylmethanol
CAS: 24324-17-2; EC No: 246-167-5; MDL No: MFCD00001139
Crystalline; Molecular Formula: C14H12O ; MW: 196.24
Melting Point: 105-107°
- Crystalline
Formula:
C₁₄H₁₂O Melting Pt: 105…107 °C |
MDL Number:
MFCD00001139 CAS Number: 24324-17-2 EINECS: 246-167-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...