6-Bromoisatin ≥95%

Supplier: BeanTown Chemical

Synonyms: 6-Bromo-1H-indole-2,3-dione, 6-Bromoindoline-2,3-dione

212995-25G 212995-5G
BT212995-25GEA 367 USD
BT212995-25G BT212995-5G
6-Bromoisatin ≥95%
6-Bromoisatin

CAS: 6326-79-0; MDL No: MFCD01631138 Solid; Molecular Formula: C8H4BrNO2; MW: 226.03 Melting Point: 274°


  • Solid

Formula: C₈H₄BrNO₂
MW: 226.03 g/mol
Melting Pt: 274 °C
MDL Number: MFCD01631138
CAS Number: 6326-79-0

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR